D(+)-Arabitol 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: D-Arabitol, (2R,4R)-Pentane-1,2,3,4,5-pentaol
- CAS Number:488-82-4
- Formula:C₅H₁₂O₅
- MDL Number:MFCD00004709
- Molecular Weight:152.15 g/mol
- Melting Pt:97…105 °C
- Storage Temperature:Refrigerator
- EINECS:207-686-2
3 Options Available Below
Product Details & Documents
Synonyms: D-Arabitol, (2R,4R)-Pentane-1,2,3,4,5-pentaol
- CAS Number:488-82-4
- Formula:C₅H₁₂O₅
- MDL Number:MFCD00004709
- Molecular Weight:152.15 g/mol
- Melting Pt:97…105 °C
- Storage Temperature:Refrigerator
- EINECS:207-686-2
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:97 to 105 °C
- HPLC:≥98.5 % (ELSD)
- Specific optical rotation:+10.5° to +13.5° (20 °C, 589 nm) (c=2, 2 % borax)



