Order Entry
Belgium
ContactUsLinkComponent
Estradiol
Estradiol
Catalog # EHERC13213100
Supplier:  EHRENSTORFER
CAS Number:  
Estradiol
Catalog # EHERC13213100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :272.39 g/mol
EINECS:200-023-8
CAS Number:50-28-2
Formula:C₁₈H₂₄O₂
MDL Number:MFCD00003693