Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hexadecacarbonylhexarhodium
Hexadecacarbonylhexarhodium ≥98%
Hexadecacarbonylhexarhodium ≥98%
STRE45-0820-250MG
:
Strem Chemicals, Inc.
:
Hexadecacarbonylhexarhodium ≥98%
STRE45-0820-250MG
:
Strem Chemicals, Inc.
:
Pack type:
Glass bottle for solids
Pk:
250 mg
Seq:
2590
:
:
1065.595 g/mol
:
2,87
:
249-009-3
:
28407-51-4
:
C₁₆O₁₆Rh₆
:
MFCD00011206