Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dodecacarbonyltetrarhodium
Dodecacarbonyltetrarhodium ≥98%
Dodecacarbonyltetrarhodium ≥98%
STRE45-2150-0250MG
:
Strem Chemicals, Inc.
:
Dodecacarbonyltetrarhodium ≥98%
STRE45-2150-0250MG
:
Strem Chemicals, Inc.
:
Pack type:
Glass bottle for solids
Pk:
250 mg
Seq:
2590
:
:
747.743 g/mol
:
2,52
:
243-171-9
:
19584-30-6
:
C₁₂O₁₂Rh₄
:
MFCD00016310