Dodecacarbonyltetrairidium ≥98%
Catalog # STRE77-1800-1G
Supplier: Strem Chemicals, Inc.
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:1104.99 g/mol
Melting Pt:230 °C (dec.)
UN:3281
ADR:6.1,I
CAS Number:11065-24-0
Formula:C₁₂Ir₄O₁₂
MDL Number:MFCD00011064