Overhead shakers, Trayster basic and digital
About this item
Overhead shakers with vertical rotation for smooth yet effective mixing, the basic model has fixed speed, and the digital unit features infinitely adjustable speed. Ideal for biological samples, particularly blood. Suitable for separating and mixing processes of powdered and liquid samples in tubes up to 50 ml. Easily removable attachments provide for quick cleaning in case of sample spills. Different attachments enable units to be used for a variety of applications.
- Can be used at 4 to 50 °C in an incubator or cold room
- Easy operation via touch keypad
- Suitable for continuous operation
- Timer: Countdown, adjustable from 1 s to 99 h 59 min (digital model)
- Digital unit has a counter: Shows shaking time and error code display
Ordering information: Supplied without attachments, required accessories must be ordered separately.
2 Options Available Below
Product Details & Documents
Overhead shakers with vertical rotation for smooth yet effective mixing, the basic model has fixed speed, and the digital unit features infinitely adjustable speed. Ideal for biological samples, particularly blood. Suitable for separating and mixing processes of powdered and liquid samples in tubes up to 50 ml. Easily removable attachments provide for quick cleaning in case of sample spills. Different attachments enable units to be used for a variety of applications.
- Can be used at 4 to 50 °C in an incubator or cold room
- Easy operation via touch keypad
- Suitable for continuous operation
- Timer: Countdown, adjustable from 1 s to 99 h 59 min (digital model)
- Digital unit has a counter: Shows shaking time and error code display
Ordering information: Supplied without attachments, required accessories must be ordered separately.
Specifications for Product Family
Specifications
- Catalog No:
- 444-1037
- 444-1038
- Model:
- Trayster basic
- Trayster digital
- Shaking motion:
- Overhead, rotating
- Overhead, rotating
- Speed:
- 20 min⁻¹ (fixed)
- 5 - 80 min⁻¹
- Max. load:
- 2 kg
- 2 kg
- Power input:
- 16 W
- 16 W
- W×D×H:
- 460×160×150 mm
- 460×160×150 mm
- Weight:
- 1,7 kg
- 1,7 kg
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- 444-1037Trayster basic overhead shakerSpecifications:
More
Specifications
Cat. No.444-1037ModelTrayster basicShaking motionOverhead, rotatingSpeed20 min⁻¹ (fixed)Max. load2 kgPower input16 WW×D×H460×160×150 mmWeight1,7 kg - 444-1038Trayster digital overhead shakerSpecifications:
More
Specifications
Cat. No.444-1038ModelTrayster digitalShaking motionOverhead, rotatingSpeed5 - 80 min⁻¹Max. load2 kgPower input16 WW×D×H460×160×150 mmWeight1,7 kg
Accessories
Product Information
- Catalog #AvailabilityPrice
- Tube insert
Product Information
- Catalog #AvailabilityPrice
- Tube insert
Product Information
- Catalog #AvailabilityPrice
- Tube insert
Product Information
- Catalog #AvailabilityPrice
- Tube insert
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



