Orders LinkContactUsLinkComponent
Microtiter tubes system
Microtiter tubes system
Microtiter tubes system
Tubes made of PP, natual, caps made of PE, natural.
  • Robotic compatible rack can easily integrated into automated liquid handling workstations
  • Polyethylene plug strip seals the microtiter tubes tightly for storage
  • Eight plug strips can be easily cut apart for individual samples
Microtiter tubes have additional volume capacity for sample dilution and are ideal for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control and blood bank sample freezer storage. Frosted writing areas on each tube and an alphanumeric rack simplify organization and identification of specific samples.
 : Non sterile: certified DNA, RNase/DNase, pyrogen and endotoxin-free.
Sterile: certified DNA, RNase/DNase, pyrogen, endotoxin, PCR inhibitor, and bioburden-free.
  • Catalog No:
  • MLBP3492
  • MLBP3487
  • MLBP3496
  • MLBP3425-11
  • MLBP3426-11
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Product Family Options
Microtiter tubes
  • Description
    Capacity
    Colour
    Packed
  • Microtiter tube, non sterile
    1,2 ml
    Natural
    Bulk
    1 * 1.000 ST
    1 * 1.000 ST - 54,00 €
    54,00 €
    1 * 1.000 ST
       54,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    MLBP3492
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
Microtiter tubes in robotic racks
  • Description
    Capacity
    Colour
    Packed
  • Microtiter tube, robotic rack, sterile
    1,2 ml
    Natural
    10 trays of 96/pack
    1 * 960 ST
    1 * 960 ST - 64,90 €
    64,90 €
    1 * 960 ST
       64,90 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    MLBP3487
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • Microtiter tube, robotic rack, non sterile
    1,2 ml
    Natural
    10 trays of 96/pack
    1 * 960 ST
    1 * 960 ST - 61,30 €
    61,30 €
    1 * 960 ST
       61,30 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    MLBP3496
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
Cap strips
  • Description
    Capacity
    Colour
    Packed
  • 8 strip pre-sterilised cap for mini tubes, non autoclavable
    Natural
    10 bags of twelve 8 cap strips
    1 * 120 ST
    1 * 120 ST - 29,30 €
    29,30 €
    1 * 120 ST
       29,30 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    MLBP3425-11
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 8 strip non-sterile cap for mini tubes, non autoclavable
    Natural
    10 bags of twelve 8 cap strips
    1 * 120 ST
    1 * 120 ST - 25,60 €
    25,60 €
    1 * 120 ST
       25,60 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    MLBP3426-11
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact