Order Entry
Belgium
ContactUsLinkComponent
Reaction vessels, cylindrical, with flat flange
Reaction vessels, cylindrical, with flat flange
N° de catalogue WITG0.900.100
Fournisseur:  witeg Labortechnik
undefined
Reaction vessels, cylindrical, with flat flange
N° de catalogue WITG0.900.100
Fournisseur:  witeg Labortechnik

Spécifications

  • Ext.Ø:
    100 mm
  • With:
    flange DN 60
  • Capacity:
    100 ml
  • Height:
    85 mm
  • Pk:
    1

Spécifications

À propos de cet article

Borosilicate glass.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

With graduation, flange with groove and round bottom.