Orders LinkContactUsLinkComponent
GC capillary column tools
GC capillary column tools
GC capillary column tools
Certificates
GC capillary column tools
2 Options Available Below
Product Details & Documents
The capillary cutting tool is used for burr free cutting of fused silica tubing.
  • Reduces likelihood of blockages
  • Ensures square ends for 'Press fit' style connectors
  • Clean entry for on-column injection
Product Family Options
Capillary washing/coating reservoir
Product Information
  • Availability
    Price
  • 558-0190
    Replacement PTFE 1/16" sealing ring
    Replacement PTFE 1/16" sealing ring
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 53,20 €
    53,20 €
    1 * 20 ST
       53,20 €
Capillary cutting tool
Product Information
  • Availability
    Price
  • 568-0470
    Capillary ceramic tube cutter
    Capillary ceramic tube cutter
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 25,10 €
    25,10 €
    1 * 3 ST
       25,10 €