ContactUsLinkComponent
Multipurpose trays, SP Bel-Art
Multipurpose trays, SP Bel-Art
Catalog # BELAF162000011
undefined
Multipurpose trays, SP Bel-Art
Catalog # BELAF162000011
Specifications
  • Capacity:
    8,5 l
  • Colour:
    Blue
  • Description:
    Multipurpose tray, with handle
  • W×D×H:
    324×267×108 mm
  • Pk:
    1
Specifications
About this item
Trays made of PP.
  • Stackable
  • Top rim adds rigidity and provides handholds for transport
  • Rounded inside corners make cleaning easy
Ideal for holding instruments while autoclaving and cleaning glassware to reduce chance of damage.