Orders LinkContactUsLinkComponent
Funnels, powder
Funnels, powder
Supplier:   Kavalier
Funnels, powder
Supplier:  Kavalier
Certificates
Funnels, powder
3 Options Available Below
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These funnels with short stem and bevelled edges are suitable for transferring powdery materials.
Ordering information: Ø = internal stem diameter.
Product Family Options
Product Information
  • Funnel O-Ø
    Stem length
    Ø
    Availability
    Price
  • 100 mm
    35 mm
    15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,30 €
    44,30 €
    1 * 1 ST
       44,30 €
  • 60 mm
    30 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,70 €
    41,70 €
    1 * 1 ST
       41,70 €
  • 80 mm
    32 mm
    12 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,60 €
    45,60 €
    1 * 1 ST
       45,60 €