Orders LinkContactUsLinkComponent
Isolator Cleaning Tools
Isolator Cleaning Tools
   Sustainable Options Available
Supplier:   MICRONOVA
Isolator Cleaning Tools
Supplier:  MICRONOVA
   Sustainable Options Available
Certificates
Isolator Cleaning Tools
16 Options Available Below
Product Details & Documents
The Isolator Tool™ adapter with a 360° swivel works with a range of mop covers to efficiently clean and disinfect isolators, hoods, RABS, and small equipment in clean and aseptic areas.
  • Low-profile design and angled head adapter
  • Available with stainless steel or plastic plates
  • PolySorb™: 100% textured polyester for additional cleaning as well as sanitisation
  • NovaPoly™: 100% smooth knit polyester provides excellent disinfectant dispersal
  • MegaTex®: Polyamide, slightly abrasive yet absorbent
  • Microfibre: 80% Polyester/20% Nylon for maximum absorbency and residue removal
  • Optional urethane foam layer to hold disinfectant and facilitate wringing
  • Compatible with alcohol and strong disinfectants
  • Launderable for reuse
  • Autoclavable
Designed for use with the SSU series Universal Handles or STA series threaded handles. Use modular and extendable handles to fit through small passes and allow extended reach.
Handles: Straight handles are available in electropolished stainless steel or plastic. Accordion handles are designed to deal with the unique space limitations in isolators and containment hoods. Available in 330 and 483 mm sizes, each section features a delrin hinge and quick disconnect to allow for attachment of additional sections or isolator tool itself.
Ordering information: Fabric covers, adapters, and handles are ordered separately. The all plastic Isolator Tool™ and slip covers are available gamma irradiated, contact your Avantor Sales Representative for more information.
Specifications for Product Family
Specifications
  • Catalog No:
  • MICRITC620-911IR
  • MICRITCAA-9
  • MICRITCAW-9
  • 129-0611
  • MICRITC6-911NSIR
  • MICRITC6-9NSIR
  • MICRITC11-9NSBB
  • MICRITC11-9NSIR
  • MICRITC11-9NS
  • 129-0625
  • 216-1490
  • MICRITC6-9
  • MICRQDAH-08
  • 129-0622
  • 129-0623
  • 141-0897
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
Specifications
Product Family Options
Covers
Product Information
  • Colour
    Length
    Availability
    Price
  • MICRITC620-911IR
    Isolator cleaning tool, MegaTex® cover with thin foam interior, irradiated, 229 mm width
    Isolator cleaning tool, MegaTex® cover with thin foam interior, irradiated, 229 mm width
    White
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 894,00 €
    894,00 €
    1 * 50 ST
       894,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRITC620-911IR
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • MICRITCAA-9
    Isolator cleaning tool, microfiber cover, 229 mm
    Isolator cleaning tool, microfiber cover, 229 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 877,00 €
    877,00 €
    1 * 50 ST
       877,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRITCAA-9
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • MICRITCAW-9
    Isolator cleaning tool, microfiber cover, 229 mm
    Isolator cleaning tool, microfiber cover, 229 mm
    White
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 877,00 €
    877,00 €
    1 * 50 ST
       877,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRITCAW-9
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 129-0611
    Isolator cleaning tool, NovaPoly cover, irradiated, 229 mm width
    Isolator cleaning tool, NovaPoly cover, irradiated, 229 mm width
    White
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 669,00 €
    669,00 €
    1 * 50 ST
       669,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    129-0611
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • MICRITC6-911NSIR
    Isolator cleaning tool, NovaPoly cover, urethane foam interior, irradiated, 229 mm width
    Isolator cleaning tool, NovaPoly cover, urethane foam interior, irradiated, 229 mm width
    White
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 606,00 €
    606,00 €
    1 * 50 ST
       606,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRITC6-911NSIR
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • MICRITC6-9NSIR
    Isolator cleaning tool, NovaPoly™ cover, irradiated, 229 mm width
    Isolator cleaning tool, NovaPoly™ cover, irradiated, 229 mm width
    White
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 481,00 €
    481,00 €
    1 * 50 ST
       481,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRITC6-9NSIR
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • MICRITC11-9NSBB
    Isolator cleaning tool, PolySorb cover, 230 mm
    Isolator cleaning tool, PolySorb cover, 230 mm
    White
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 476,00 €
    476,00 €
    1 * 50 ST
       476,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MICRITC11-9NSBB
    Environmentally Preferable
  • MICRITC11-9NSIR
    Isolator cleaning tool, PolySorb cover, irradiated, 230 mm
    Isolator cleaning tool, PolySorb cover, irradiated, 230 mm
    White
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 560,00 €
    560,00 €
    1 * 50 ST
       560,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRITC11-9NSIR
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • MICRITC11-9NS
    Isolator cleaning tool, PolySorb™ cover, 229 mm width
    Isolator cleaning tool, PolySorb™ cover, 229 mm width
    White
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 476,00 €
    476,00 €
    1 * 50 ST
       476,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRITC11-9NS
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • MICRITC6-9
    NovaPoly™ 100% smooth knit polyester cover, 23cm length, elastic band attachment
    NovaPoly™ 100% smooth knit polyester cover, 23cm length, elastic band attachment
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 554,00 €
    554,00 €
    1 * 50 ST
       554,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRITC6-9
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
Isolator tools
Product Information
  • Colour
    Length
    Availability
    Price
  • 129-0625
    Isolator tool adapter, 229 mm
    Isolator tool adapter, 229 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 253,00 €
    253,00 €
    1 * 1 ST
       253,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    129-0625
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 216-1490
    Isolator tool adapter, plastic, 229 mm
    Isolator tool adapter, plastic, 229 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 115,00 €
    115,00 €
    1 * 1 ST
       115,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    216-1490
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
Handles
Product Information
  • Colour
    Length
    Availability
    Price
  • MICRQDAH-08
    Over/under accordion handle
    Over/under accordion handle
    203 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 274,00 €
    274,00 €
    1 * 1 ST
       274,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRQDAH-08
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 129-0622
    Over/under accordion handle
    Over/under accordion handle
    330 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 274,00 €
    274,00 €
    1 * 1 ST
       274,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    129-0622
    Environmentally Preferable
  • 129-0623
    Over/under accordion handle
    Over/under accordion handle
    483 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 290,00 €
    290,00 €
    1 * 1 ST
       290,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    129-0623
    Environmentally Preferable
  • 141-0897
    Plastic ACME threaded extendable handle, with white hand grip
    Plastic ACME threaded extendable handle, with white hand grip
    305 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 26,80 €
    26,80 €
    1 * 1 Pce
       26,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    141-0897
    Environmentally Preferable