Orders LinkContactUsLinkComponent
Flathead mop covers, SnapMop™
Flathead mop covers, SnapMop™
   Sustainable Options Available
Supplier:   MICRONOVA
Flathead mop covers, SnapMop™
Supplier:  MICRONOVA
   Sustainable Options Available
Certificates
Flathead mop covers, SnapMop™
16 Options Available Below
Product Details & Documents
A range of lightweight, snap-on flat head mop covers with urethane foam interiors, designed for effective cleaning and sanitising in the cleanroom. A variety of fabric options provide excellent absorbency, effective cleaning, and disinfectant dispersal for specialised cleaning.
  • Easy to change covers fit securely over mop frame
  • Urethane foam interiors fully encased in a choice of fabrics
  • NovaPoly: 100% smooth knit polyester provides excellent disinfectant dispersal
  • PolySorb: 100% textured polyester for removing residues and dispersing disinfectant
  • MegaTex: 100% non-woven polyamide is absorbent with a mildly abrasive surface
  • PolyMesh: 100% polyester with a layer of mesh provides a mild scrubbing surface
  • 228,6 mm covers are excellent for pass-throughs and smaller spaces
  • 355,6 and 457,2 mm covers are ideal for floors, walls, and ceilings
Covers are constructed with a thin urethane foam interior completely encased in polyester, ensuring no exposed foam and resulting in less weight and faster drying. Available in 100% polyester or with 'cleaning bands' in microfibre for additional cleaning power or MegaTex® for cleaning rough/textured surfaces or removing heavy deposits. Can be laundered, re-used and able to withstand alcohol, strong disinfectants and steam sterilisation. Also available gamma irradiated.
Stainless steel snap fasteners connection.
Specifications for Product Family
Specifications
  • Catalog No:
  • MICRMZSM11-9NS
  • MICRMZSM11-14NS
  • 141-1021
  • 141-1022
  • 141-1026
  • 129-0974
  • 141-1023
  • 141-1024
  • 141-1025
  • 141-1031
  • 141-1032
  • 129-0975
  • 141-1029
  • 141-1030
  • MICRMZSM6AW-14
  • MICRMZSM6AW-18
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
Specifications
Product Family Options
PolySorb™ SnapMop™ Flat head mop covers
Product Information
  • Length
    Availability
    Price
  • MICRMZSM11-9NS
    SnapMop™ Flat head mop covers; no sponge interior; no snaps, PolySorb™, 100% textured polyester (without foam)
    SnapMop™ Flat head mop covers; no sponge interior; no snaps, PolySorb™, 100% textured polyester (without foam)
    230 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 743,00 €
    743,00 €
    1 * 50 ST
       743,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRMZSM11-9NS
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • MICRMZSM11-14NS
    SnapMop™ Flat head mop covers; no sponge interior; no snaps, PolySorb™, 100% textured polyester (without foam)
    SnapMop™ Flat head mop covers; no sponge interior; no snaps, PolySorb™, 100% textured polyester (without foam)
    360 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 759,00 €
    759,00 €
    1 * 50 ST
       759,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRMZSM11-14NS
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
NovaPoly™ SnapMop™ Flat head mop covers
Product Information
  • Length
    Availability
    Price
  • 141-1021
    SnapMop™ Flat head mop covers, NovaPoly™, 100% smooth knit polyester
    SnapMop™ Flat head mop covers, NovaPoly™, 100% smooth knit polyester
    230 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 518,00 €
    518,00 €
    1 * 24 ST
       518,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    141-1021
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 141-1022
    Irradiated SnapMop™ Flat head mop covers, NovaPoly™, 100% smooth knit polyester
    Irradiated SnapMop™ Flat head mop covers, NovaPoly™, 100% smooth knit polyester
    230 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 705,00 €
    705,00 €
    1 * 24 ST
       705,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    141-1022
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
MegaTex® SnapMop™ Flat head mop covers
Product Information
  • Length
    Availability
    Price
  • 141-1026
    SnapMop™ Flat head mop covers, MegaTex®, 100% non-woven polyamide
    SnapMop™ Flat head mop covers, MegaTex®, 100% non-woven polyamide
    230 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 548,00 €
    548,00 €
    1 * 24 ST
       548,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    141-1026
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 129-0974
    SnapMop™ Flat head mop covers, MegaTex®, 100% non-woven polyamide
    SnapMop™ Flat head mop covers, MegaTex®, 100% non-woven polyamide
    360 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,20 €
    25,20 €
    1 * 1 ST
       25,20 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    129-0974
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 141-1023
    Irradiated SnapMop™ Flat head mop covers, MegaTex®, 100% non-woven polyamide
    Irradiated SnapMop™ Flat head mop covers, MegaTex®, 100% non-woven polyamide
    360 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 819,00 €
    819,00 €
    1 * 24 ST
       819,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    141-1023
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 141-1024
    SnapMop™ Flat head mop covers, MegaTex®, 100% non-woven polyamide
    SnapMop™ Flat head mop covers, MegaTex®, 100% non-woven polyamide
    460 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 629,00 €
    629,00 €
    1 * 24 ST
       629,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    141-1024
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 141-1025
    Irradiated SnapMop™ Flat head mop covers, MegaTex®, 100% non-woven polyamide
    Irradiated SnapMop™ Flat head mop covers, MegaTex®, 100% non-woven polyamide
    460 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 863,00 €
    863,00 €
    1 * 24 ST
       863,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    141-1025
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
Blue Microfiber SnapMop™ Flat head mop covers
Product Information
  • Length
    Availability
    Price
  • 141-1031
    SnapMop™ Flat head mop covers; blue, microfiber, 80% polyester; 20% polyamide
    SnapMop™ Flat head mop covers; blue, microfiber, 80% polyester; 20% polyamide
    230 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 548,00 €
    548,00 €
    1 * 24 ST
       548,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    141-1031
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 141-1032
    Irradiated SnapMop™ Flat head mop covers; blue, microfiber, 80% polyester; 20% polyamide
    Irradiated SnapMop™ Flat head mop covers; blue, microfiber, 80% polyester; 20% polyamide
    230 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 731,00 €
    731,00 €
    1 * 24 ST
       731,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    141-1032
    Environmentally Preferable
  • 129-0975
    SnapMop™ Flat head mop covers; blue, microfiber, 80% polyester; 20% polyamide
    SnapMop™ Flat head mop covers; blue, microfiber, 80% polyester; 20% polyamide
    360 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,20 €
    25,20 €
    1 * 1 ST
       25,20 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    129-0975
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 141-1029
    SnapMop™ Flat head mop covers; blue, microfiber, 80% polyester; 20% polyamide
    SnapMop™ Flat head mop covers; blue, microfiber, 80% polyester; 20% polyamide
    460 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 629,00 €
    629,00 €
    1 * 24 ST
       629,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    141-1029
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • 141-1030
    Irradiated SnapMop™ Flat head mop covers; blue, microfiber, 80% polyester; 20% polyamide
    Irradiated SnapMop™ Flat head mop covers; blue, microfiber, 80% polyester; 20% polyamide
    460 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 863,00 €
    863,00 €
    1 * 24 ST
       863,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    141-1030
    Environmentally Preferable
White Microfiber SnapMop™ Flat head mop covers
Product Information
  • Length
    Availability
    Price
  • MICRMZSM6AW-14
    SnapMop™ Flat head mop covers; white microfiber, 80% polyester; 20% polyamide
    SnapMop™ Flat head mop covers; white microfiber, 80% polyester; 20% polyamide
    360 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 603,00 €
    603,00 €
    1 * 24 ST
       603,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRMZSM6AW-14
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
  • MICRMZSM6AW-18
    SnapMop™ Flat head mop covers; white microfiber, 80% polyester; 20% polyamide
    SnapMop™ Flat head mop covers; white microfiber, 80% polyester; 20% polyamide
    460 mm
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 629,00 €
    629,00 €
    1 * 24 ST
       629,00 €
    Specifications:
      Waste Reducing
    More
    Specifications
    Cat. No.
    MICRMZSM6AW-18
    Environmentally Preferable
    Waste Reducing
      Waste Reducing
Stainless steel handles
Product Information
  • Catalog #
    Description
    Length
    Material
    Availability
    Price
  • Stainless steel quick disconnect extendable handle
    965 - 1829 mm
    Stainless steel 304
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 314,00 €
    314,00 €
    1 * 1 ST
       314,00 €
Product Information
  • Catalog #
    Description
    Length
    Material
    Availability
    Price
  • Stainless steel quick disconnect handle
    203 mm
    Stainless steel 304
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 98,70 €
    98,70 €
    1 * 1 ST
       98,70 €
Product Information
  • Catalog #
    Material
    Description
    Length
    Availability
    Price
  • 03
    Stainless steel quick disconnect extendable handle
    1575 − 3048 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 389,00 €
    389,00 €
    1 * 1 ST
       389,00 €
Product Information
  • Catalog #
    Description
    Length
    Material
    Availability
    Price
  • Stainless steel quick disconnect handle
    1295 cm
    Stainless steel 304
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 140,00 €
    140,00 €
    1 * 1 ST
       140,00 €
Plastic handles
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • Plastic ACME threaded extendable handle, with white hand grip
    102 - 183 cm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 144,00 €
    144,00 €
    1 * 1 ST
       144,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • Plastic ACME threaded handle, with white hand grip
    152 cm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,90 €
    46,90 €
    1 * 1 ST
       46,90 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • Plastic ACME threaded extendable handle, with white hand grip
    1016 - 2565 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 224,00 €
    224,00 €
    1 * 1 ST
       224,00 €
Product Information
  • Catalog #
    Description
    Length
    Availability
    Price
  • Plastic ACME threaded handle, with white hand grip
    1219 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,70 €
    42,70 €
    1 * 1 ST
       42,70 €
Accessories
Product Information
  • Catalog #
    L×W
    Availability
    Price
  • SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 9" flat head mops; SSU series handles
    SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 9" flat head mops; SSU series handles
    129-0536
    9×4" (22.9×10 cm) mm
    Log in for availability
    1 * 1 ST - 333,00 €
Product Information
  • Catalog #
    Availability
    Price
  • SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 14" flat head mops; SSU series handles, 14×4" (35.5×10 cm)
    SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 14" flat head mops; SSU series handles, 14×4" (35.5×10 cm)
    129-0537
    Log in for availability
    1 * 1 ST - 379,00 €
Product Information
  • Catalog #
    L×W
    Availability
    Price
  • SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 18" flat head mops; SSU series handles
    SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 18" flat head mops; SSU series handles
    129-0538
    18×4" (45.7×10 cm) mm
    Log in for availability
    1 * 1 ST - 395,00 €
Product Information
  • Catalog #
    L×W
    Availability
    Price
  • SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 9" flat head mops; STA series handles , 9" flat head mops; STA series handles
    SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 9" flat head mops; STA series handles , 9" flat head mops; STA series handles
    MICRQPSL-09
    9×4" (22.9×10 cm) mm
    Log in for availability
    1 * 1 ST - 132,00 €
Product Information
  • Catalog #
    L×W
    Availability
    Price
  • SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 14" flat head mops; STA series handles
    SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 14" flat head mops; STA series handles
    129-0976
    14×4" (35.5×10 cm) mm
    Log in for availability
    1 * 1 ST - 144,00 €
Product Information
  • Catalog #
    L×W
    Availability
    Price
  • SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 18" flat head mops; STA series handles
    SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 18" flat head mops; STA series handles
    MICRQPSL-18
    18×4" (45.7×10 cm) mm
    Log in for availability
    1 * 1 ST - 158,00 €