Orders LinkContactUsLinkComponent
Digital refractometers, PR-α series
Supplier:   Atago
Digital refractometers, PR-α series
Supplier:  Atago
Certificates
Digital refractometers, PR-α series
4 Options Available Below
Product Details & Documents
Modern, attractive refractometers for sugar applications.
  • Different colours to identify the models
  • Simple two-button operation
  • Calibration with water
External light interference (ELI) to take accurate measurements in daylight or near a window.
 : PR-301α is supplied with a calibration standard (7 ml) and round sheets.
Specifications for Product Family
Specifications
  • Catalog No:
  • 635-0152
  • 635-0151
  • 635-0149
  • 635-0150
  • Temperature compensation range:
  • 5/10...40 °C
  • 5/10...40 °C
  • 5/10...40 °C
  • 5/10...40 °C
  • Electrical:
  • 9 V 006p dry battery
  • 9 V 006p dry battery
  • 9 V 006p dry battery
  • 9 V 006p dry battery
  • W×D×H:
  • 170×90×40 mm
  • 170×90×40 mm
  • 170×90×40 mm
  • 170×90×40 mm
  • Weight:
  • 300 g
  • 300 g
  • 300 g
  • 300 g
Specifications
Product Family Options
Product Information
  • Model
    Range
    Resolution
    Availability
    Price
  • PR-301α
    45,0 - 90,0 °Brix
    0,1 °Brix
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 390,00 €
    1 390,00 €
    1 * 1 ST
       1 390,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    635-0152
    Temperature compensation range
    5/10...40 °C
    Electrical
    9 V 006p dry battery
    W×D×H
    170×90×40 mm
    Weight
    300 g
  • PR-201α
    0,0 - 60,0 °Brix
    0,1 °Brix
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 640,00 €
    1 640,00 €
    1 * 1 ST
       1 640,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    635-0151
    Temperature compensation range
    5/10...40 °C
    Electrical
    9 V 006p dry battery
    W×D×H
    170×90×40 mm
    Weight
    300 g
  • 635-0149
    Digital refractometer
    Digital refractometer
    PR-32α
    0,0 - 32,0 °Bx
    0,1 °Bx
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 629,00 €
    629,00 €
    1 * 1 ST
       629,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    635-0149
    Temperature compensation range
    5/10...40 °C
    Electrical
    9 V 006p dry battery
    W×D×H
    170×90×40 mm
    Weight
    300 g
  • 635-0150
    Digital refractometer
    Digital refractometer
    PR-101α
    0,0 - 45,0 °Brix
    0,1 °Brix
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 964,00 €
    964,00 €
    1 * 1 ST
       964,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    635-0150
    Temperature compensation range
    5/10...40 °C
    Electrical
    9 V 006p dry battery
    W×D×H
    170×90×40 mm
    Weight
    300 g