ContactUsLinkComponent
Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
  SEMA2025
 :  SEMADENI
undefined
Packaging bottles, narrow neck, rectangular, HDPE, with screw caps
  SEMA2025
 :  SEMADENI
  • Capacity:
    500 ml
  • W×D×H:
    60×70×160 mm
  • Neck I-Ø:
    22 mm
  • Pk:
    100
HDPE, translucent, with thread.
  • Good chemical resistance
  • Good temperature resistance
 : Screw caps must be ordered separately.
Screw caps and sealing caps