Order Entry
Belgium
ContactUsLinkComponent
 
Sodium hydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
 :  Avantor
 
undefined
Sodium hydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
 :  Avantor
 

 

  • For Laboratory,Research,or Manufacturing Use:
    50 - 52%
  • Protect from air to avoid absorption of carbon dioxide.:
    ≤20
  • Assay (NaOH) (by acidimetry):
    50 - 52 %
  • Color (APHA):
    ≤20
  • Sodium Carbonate (Na₂CO₃):
    ≤0.1 %
  • Chloride (Cl):
    ≤0.002 %
  • Sulfate (SO₄):
    ≤0.001 %
  • Ammonium Hydroxide Precipitate:
    ≤0.01 %
  • Silica (SiO₂), For Information Only:
    ≤5 ppm
  • Heavy Metals (as Ag):
    ≤0.001 %
  • Potassium (K):
    ≤0.01 %
  • Nitrogen Compounds (as N):
    ≤5 ppm
  • Phosphate (PO₄):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm
  • Trace Impurities - Nickel (Ni):
    ≤5.000 ppm

 

Frequently Bought Together