L-Threonine ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:72-19-5
:
Formula:C₄H₉NO₃
MDL Number:MFCD00064270
:119.12 g/mol
Merck Index:14,09380
Storage Temperature:Ambient
EINECS:200-774-1
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 %
- Specific rotation [a]20/D:-27.5 to -29.0 deg (C=6, H2O)
- Solubility in Water:almost transparency
Specification Test Results
Related Information
Frequently Bought Together