Order Entry
Belgium
Orders LinkContactUsLinkComponent
Formic acid

Formic acid

1 KIT
Catalog # 710-1895
Flammable
Corrosive
Supplier:   MEGAZYME
Formic acid
Catalog # 710-1895
Supplier:  MEGAZYME
CAS Number:  64-18-6
Formic acid
Formic acid

1 KIT

CAS Number:  64-18-6
Price
276,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Synonyms:  Aminic Acid, Methanoic acid
  • CAS Number:64-18-6
  • Formula:HCO₂H
  • MDL Number:MFCD00003297
  • Molecular Weight:46.03 g/mol
  • Boiling Pt:100…101 °C (1013 hPa)
  • Melting Pt:8.2…8.4 °C
  • Density:1.22 g/cm³ (25 °C)
  • Flash Pt:48 °C
  • EINECS:200-579-1
  • ADR:8,II
  •  :Density: 1.22 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • UN:1779
  • REACH:01-2119491174-37
Specifications

Specifications

  • Pk:
    1 KIT

Specifications