
Methyl hydrogen adipate ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:627-91-8
:
Formula:C₇H₁₂O₄
MDL Number:MFCD00004418
:160.17 g/mol
Boiling Pt:162°C
Melting Pt:9°C
Density:1,1236 g/cm³ (20 °C)
EINECS:211-019-0
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together