Order Entry
Belgium
ContactUsLinkComponent
Buffer, sodium acetate, 3 M aq, soln, pH 5,2, RNAse free
Buffer, sodium acetate, 3 M aq, soln, pH 5,2, RNAse free
Certificates
Buffer, sodium acetate, 3 M aq, soln, pH 5,2, RNAse free
Certificates

Specifications

  • CAS No.:
    127-09-3
  • MDL No.:
    MFCD00012459
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless

Specifications

Related Information

Frequently Bought Together