Sodium acetate 45% in aqueous solution, extra pure suitable as buffer solution for photometric determination of iron
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:127-09-3
:
Formula:H₃CCOONa
MDL Number:MFCD00012459
:82.03 g/mol
Storage Temperature:Ambient
EINECS:204-823-8
Frequently Bought Together