Order Entry
Belgium
ContactUsLinkComponent
Benzophenone-[D10] 100 µg/ml in n-nonane (98% D)
Benzophenone-[D10] 100 µg/ml in n-nonane (98% D)
Catalog # PROOCILDLM-183-1.2
CAS Number:  
Benzophenone-[D10] 100 µg/ml in n-nonane (98% D)
Catalog # PROOCILDLM-183-1.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    22583-75-1
  • Description:
    Benzophenone-[D10] 100 µg/ml in n-nonane (98% D)
  • Formula:
    C₁₃D₁₀O
  • Description:
    Benzophenone-[D10]

Specifications

About this item