Order Entry
Belgium
ContactUsLinkComponent
Trypan blue 0,4% in aqueous solution, Sigma-Aldrich®
Health
Trypan blue 0,4% in aqueous solution, Sigma-Aldrich®
Catalog # 93595-50ML
CAS Number:  
undefined
Health
Trypan blue 0,4% in aqueous solution, Sigma-Aldrich®
Catalog # 93595-50ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Appearance (colour):
    Blue to very dark blue
  • Appearance (form):
    Liquid
  • Thin layer chromatogr.:
    Corresponds to standard
  • Density d20/4:
    0.997 - 1.017
  • Wavelength (1) (UV):
    600 - 610 nm
  • Solvent (UV):
    Methanol
  • Expiry period:
    30 months

Specification Test Results

About this item

 :
 :960.82 g/mol
Density:1,007 g/ml at 20 °C
 :M-Clarity™ quality level = MQ200
EINECS:200-786-7
 :GHS08
Storage Temperature:Room Temperature
CAS Number:72-57-1
Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number:MFCD00003969