Order Entry
Belgium
ContactUsLinkComponent
Silicon standard dissolved in oil, 5,000 µg/g Si in mineral oil for wear metal analysis, Specpure®
Silicon standard dissolved in oil, 5,000 µg/g Si in mineral oil for wear metal analysis, Specpure®
Catalog # 43867.18
CAS Number:  
Silicon standard dissolved in oil, 5,000 µg/g Si in mineral oil for wear metal analysis, Specpure®
Catalog # 43867.18
CAS Number:  

Specifications

  • Concentration:
    5000 µg/g
  • Matrix:
    Mineral oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Description:
    Standard for wear metal analysis
  • MDL No.:
    MFCD00085311
  • Element:
    Silicon
  • Symbol:
    Si

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00085311