Order Entry
Belgium
ContactUsLinkComponent
Iron standard dissolved in oil, 5000 µg/g Fe in mineral oil for wear metal analysis, Specpure®
Iron standard dissolved in oil, 5000 µg/g Fe in mineral oil for wear metal analysis, Specpure®
Catalog # 42440.18
CAS Number:  
Iron standard dissolved in oil, 5000 µg/g Fe in mineral oil for wear metal analysis, Specpure®
Catalog # 42440.18
CAS Number:  

Specifications

  • Concentration:
    5000 mg/L
  • Matrix:
    Mineral oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • MDL No.:
    MFCD00010999
  • Element:
    Iron
  • Symbol:
    Fe

Specifications

About this item

Physical form: liquid
MDL No.: MFCD00010999