Order Entry
Belgium
ContactUsLinkComponent
Bismuth standard dissolved in oil, 1000 µg/g Bi in mineral oil for wear metal analysis, Specpure®
Bismuth standard dissolved in oil, 1000 µg/g Bi in mineral oil for wear metal analysis, Specpure®
Catalog # 14223.18
CAS Number:  
Bismuth standard dissolved in oil, 1000 µg/g Bi in mineral oil for wear metal analysis, Specpure®
Catalog # 14223.18
CAS Number:  

Specifications

  • Concentration:
    1000 µg/g
  • Matrix:
    Mineral oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • MDL No.:
    MFCD00134033
  • Element:
    Bismuth
  • Symbol:
    Bi

Specifications

About this item

Physical form: liquid
MDL No.: MFCD00134033