Order Entry
Belgium
ContactUsLinkComponent
Magnesium standard dissolved in oil, 5000 µg/g Mg in mineral oil for wear metal analysis, Specpure®
Magnesium standard dissolved in oil, 5000 µg/g Mg in mineral oil for wear metal analysis, Specpure®
Catalog # 42443.18
CAS Number:  
Magnesium standard dissolved in oil, 5000 µg/g Mg in mineral oil for wear metal analysis, Specpure®
Catalog # 42443.18
CAS Number:  

Specifications

  • Concentration:
    5000 mg/L
  • Matrix:
    Mineral oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • MDL No.:
    MFCD00085308
  • Element:
    Magnesium
  • Symbol:
    Mg

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00085308