Order Entry
Belgium
ContactUsLinkComponent
 
6-O-Palmitoyl-L-ascorbic acid 98.0-100.5%, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, FCC, E304, SAFC®
 
6-O-Palmitoyl-L-ascorbic acid 98.0-100.5%, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, FCC, E304, SAFC®
 

 

  • Assay (iodometric, calc. on dried substance):
    98.0 - 100.5 %
  • Identity (IR-spectrum):
    conforms
  • Identity (Tillmans-Reaction):
    conforms
  • Appearance:
    white or yellowish-white powder
  • Appearance of solution (100 g/l, methanol):
    clear and not more intense in colour than colour reference solution BY4 (Ph. Eur.)
  • Melting range (lower value):
    ≥ 107 °C
  • Melting range (upper value):
    ≤ 117 °C
  • Spec. rotation α 20/D (100 g/l, Metanol, calc on dried basis):
    +21 - +24 °
  • Spec. rotation α 20/D (50 g/l, methanol):
    +21 - +24 °
  • Spec. rotation α 25/D (100 g/l, Metanol, calc on dried basis):
    +21 - +24 °
  • Spec. rotation α 25/D (100 g/l, methanol):
    +21 - +24 °
  • Heavy metals (as Pb):
    ≤ 10 ppm
  • As (Arsenic):
    ≤ 3 ppm
  • Cu (Copper):
    ≤ 25 ppm
  • Hg (Mercury):
    ≤ 1 ppm
  • Pb (Lead):
    ≤ 2 ppm
  • Zn (Zinc):
    ≤ 25 ppm
  • Ethyl acetate (HS-GC):
    ≤ 5000 ppm
  • Methylisobutylketone (HS-GC):
    ≤ 5000 ppm
  • Other residual solvents (ICH Q3C):
    excluded by manufacuring process
  • Sulfated ash (600 °C):
    ≤ 0.1 %
  • Loss on drying (5 h, 60 °C, Vacuum):
    ≤ 1.0 %
  • Residues of metal catalysts or metal reagents acc.to EMEA/CHMP/SWP/4446/2000:
    are not likely to be present.
  • Correspond to Ph Eur, BP, NF, FCC, E 304:
    Conforms to the purity criteria on food additives according to the current ECR

 

Frequently Bought Together