Order Entry
Belgium
ContactUsLinkComponent
Barium standard dissolved in oil, 5,000 µg/g Ba in mineral oil for wear metal analysis, Specpure®
Barium standard dissolved in oil, 5,000 µg/g Ba in mineral oil for wear metal analysis, Specpure®
Catalog # 42434.18
CAS Number:  
Barium standard dissolved in oil, 5,000 µg/g Ba in mineral oil for wear metal analysis, Specpure®
Catalog # 42434.18
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Mineral oil
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • MDL No.:
    MFCD00134031
  • Element:
    Barium
  • Symbol:
    Ba

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00134031