Theophyllin monohydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5967-84-0
:
Formula:C₇H₈N₄O₂·1H₂O
MDL Number:MFCD00151659
:198.18 g/mol (180.16 anhy)
Merck Index:14,9285
Melting Pt:270…274 °C
Storage Temperature:Ambient
EINECS:200-385-7
UN:2811
ADR:6.1,III
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together