
2-Sulphobenzoic cyclic anhydride ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:81-08-3
:
Formula:C₇H₄O₄S
MDL Number:MFCD00005879
:184.17 g/mol
EINECS:201-322-6
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Methanolysis Method):min. 97.0 %
- Melting point:123.0 to 128.0 deg-C
Specification Test Results
Frequently Bought Together