Order Entry
Belgium
Orders LinkContactUsLinkComponent
D(-)-Luciferin

D(-)-Luciferin

Supplier:   Serva
D(-)-Luciferin
Supplier:  Serva
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
  • CAS Number:2591-17-5
  • Formula:C₁₁H₈N₂O₃S₂
  • MDL Number:MFCD00042929
  • Molecular Weight:280.3228 g/mol
  • EINECS:219-981-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    2 mg
    Log in for availability
    Log in for availability
    1 * 2 mg
    1 * 2 mg

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg

    Specifications:
    Marketsource
    Marketsource
  • 250 mg
    Discontinued
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.