Order Entry
Belgium
Orders LinkContactUsLinkComponent
D(-)-Luciferin

D(-)-Luciferin

Supplier:   Biotium
D(-)-Luciferin
Supplier:  Biotium
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
  • CAS Number:2591-17-5
  • Formula:C₁₁H₈N₂O₃S₂
  • MDL Number:MFCD00042929
  • Molecular Weight:280.3228 g/mol
  • EINECS:219-981-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 135,00 €
    135,00 €
    1 * 10 mg
       135,00 €

  • Glass bottle for solids
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 388,00 €
    388,00 €
    1 * 50 mg
       388,00 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 2 330,00 €
    2 330,00 €
    1 * 1 g
       2 330,00 €