Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, TRIS, 1,0 M Solution, pH 9,0

Buffer, TRIS, 1,0 M Solution, pH 9,0

TRIS buffer solution pH 9 (1 mol/L), Glass bottle, 1 L
Catalog # J62085.K2
Buffer, TRIS, 1,0 M Solution, pH 9,0
Catalog # J62085.K2
CAS Number:  
Buffer, TRIS, 1,0 M Solution, pH 9,0

TRIS buffer solution pH 9 (1 mol/L), Glass bottle, 1 L

CAS Number:  
Price
112,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TRIS buffer solution pH 9 (1 mol/L)
  • Cat. No.:
    J62085.K2
  • Odour/scent:
    Odourless
  • Storage temperature:
    Ambient
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00004679

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.