Order Entry
Belgium
ContactUsLinkComponent
Buffers, TRIS, 1,0 M solutions, 0,2 µm filtered
Buffers, TRIS, 1,0 M solutions, 0,2 µm filtered
Certificates
Buffers, TRIS, 1,0 M solutions, 0,2 µm filtered
Certificates

Specifications

  • CAS No.:
    77-86-1
  • MDL No.:
    MFCD00004679
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless

Specifications

Frequently Bought Together