Order Entry
Belgium
ContactUsLinkComponent
 
Buffer, tris(hydroxymethyl)aminomethane hydrochloride, 1 M stock soln, pH 8,0
  J63636.K2
 :  
Buffer, tris(hydroxymethyl)aminomethane hydrochloride, 1 M stock soln, pH 8,0
  J63636.K2
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • CAS No.:
    1185-53-1
  • Description:
    TRIS buffer solution pH 8 (1 mol/l)
  • MDL No.:
    MFCD00012590
  • Physical form:
    Liquid
  • Appearance:
    Colourless

 

 

CAS No.: 1185-53-1
Physical form: liquid
Appearance: colourless
MDL No.: MFCD00012590