Order Entry
Belgium
ContactUsLinkComponent
Buffer, tris(hydroxymethyl)aminomethane hydrochloride, 1 M solution, pH 8,0, RNase-free
Buffer, tris(hydroxymethyl)aminomethane hydrochloride, 1 M solution, pH 8,0, RNase-free
Catalog # J60080.AK
CAS Number:  
Buffer, tris(hydroxymethyl)aminomethane hydrochloride, 1 M solution, pH 8,0, RNase-free
Catalog # J60080.AK
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • CAS No.:
    1185-53-1
  • Description:
    TRIS buffer solution pH 8 (1 mol/l)
  • MDL No.:
    MFCD00012590
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

About this item

CAS No.: 1185-53-1
Physical form: liquid
Appearance: colourless
MDL No.: MFCD00012590