Order Entry
Belgium
ContactUsLinkComponent
Buffers, PIPES, 1,0 M solutions
Buffers, PIPES, 1,0 M solutions
Certificates
Buffers, PIPES, 1,0 M solutions
Certificates

Specifications

  • CAS No.:
    5625-37-6
  • MDL No.:
    MFCD00006159
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

Frequently Bought Together