Order Entry
Belgium
ContactUsLinkComponent
Buffer, sodium acetate, 1M aq, soln,, pH 4,5, RNAse free
Buffer, sodium acetate, 1M aq, soln,, pH 4,5, RNAse free
Catalog # J63669.AK
CAS Number:  
Buffer, sodium acetate, 1M aq, soln,, pH 4,5, RNAse free
Catalog # J63669.AK
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • CAS No.:
    127-09-3
  • Description:
    Sodium acetate, 1M aq, solution, pH 4,5, RNAse free
  • MDL No.:
    MFCD00012459
  • Formula weight:
    82.03
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless
  • Molecular formula:
    C2H3NaO2

Specifications

About this item