Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

HEPES, 1,0 M buffer solutions

HEPES, 1,0 M buffer solutions
Certificates
HEPES, 1,0 M buffer solutions

12 Options Available Below

Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • J61017.AK
  • J61017.AP
  • J62688.AK
  • J62688.AP
  • J60712.AK
  • J60712.AP
  • J63578.AK
  • J63578.AP
  • J61360.AK
  • J61360.AP
  • J60440.AK
  • J60440.AP
  • Physical form:
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • CAS No.:
  • 7365-45-9
  • 7365-45-9
  • 7365-45-9
  • 7365-45-9
  • 7365-45-9
  • 7365-45-9
  • 7365-45-9
  • 7365-45-9
  • 7365-45-9
  • 7365-45-9
  • MDL No.:
  • MFCD00006158
  • MFCD00006158
  • MFCD00006158
  • MFCD00006158
  • MFCD00006158
  • MFCD00006158
  • MFCD00006158
  • MFCD00006158
  • MFCD00006158
  • MFCD00006158
  • Appearance:
  • Colourless
  • Colourless
  • Storage temperature:
  • 2°C to 8°C
  • 2°C to 8°C
  • 2°C to 8°C
  • 2°C to 8°C

Specifications

Product Family Options

Product Information

  • Concentration
    pH value
    Pack type
    Pk
    Availability
    Price
  • J61017.AK
    HEPES buffer solution
    HEPES buffer solution
    1 mol/L
    6,5
    Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 163,00 €
    163,00 €
    1 * 250 mL
       163,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61017.AK
    Physical form
    Liquid
    CAS No.
    7365-45-9
    MDL No.
    MFCD00006158
    Appearance
    Storage temperature
  • J61017.AP
    HEPES buffer solution
    HEPES buffer solution
    1 mol/L
    6,5
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 236,00 €
    236,00 €
    1 * 500 mL
       236,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61017.AP
    Physical form
    Liquid
    CAS No.
    7365-45-9
    MDL No.
    MFCD00006158
    Appearance
    Storage temperature
  • J62688.AK
    HEPES buffer solution
    HEPES buffer solution
    1 mol/L
    7,0
    Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 151,00 €
    151,00 €
    1 * 250 mL
       151,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J62688.AK
    Physical form
    Liquid
    CAS No.
    7365-45-9
    MDL No.
    MFCD00006158
    Appearance
    Colourless
    Storage temperature
    2°C to 8°C
  • J62688.AP
    HEPES buffer solution
    HEPES buffer solution
    1 mol/L
    7,0
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 228,00 €
    228,00 €
    1 * 500 mL
       228,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J62688.AP
    Physical form
    Liquid
    CAS No.
    7365-45-9
    MDL No.
    MFCD00006158
    Appearance
    Colourless
    Storage temperature
    2°C to 8°C
  • J60712.AK
    HEPES buffer solution
    HEPES buffer solution
    1 mol/L
    7,5
    Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 177,00 €
    177,00 €
    1 * 250 mL
       177,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60712.AK
    Physical form
    Liquid
    CAS No.
    7365-45-9
    MDL No.
    MFCD00006158
    Appearance
    Storage temperature
    2°C to 8°C
  • J60712.AP
    HEPES buffer solution
    HEPES buffer solution
    1 mol/L
    7,5
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 248,00 €
    248,00 €
    1 * 500 mL
       248,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60712.AP
    Physical form
    Liquid
    CAS No.
    7365-45-9
    MDL No.
    MFCD00006158
    Appearance
    Storage temperature
    2°C to 8°C
  • J63578.AK
    HEPES buffer solution
    HEPES buffer solution
    1 mol/L
    8,0
    Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 175,00 €
    175,00 €
    1 * 250 mL
       175,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J63578.AK
    Physical form
    Liquid
    CAS No.
    7365-45-9
    MDL No.
    MFCD00006158
    Appearance
    Storage temperature
  • J63578.AP
    HEPES buffer solution
    HEPES buffer solution
    1 mol/L
    8,0
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 236,00 €
    236,00 €
    1 * 500 mL
       236,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J63578.AP
    Physical form
    Liquid
    CAS No.
    7365-45-9
    MDL No.
    MFCD00006158
    Appearance
    Storage temperature
  • J61360.AK
    HEPES buffer solution
    HEPES buffer solution
    1 mol/L
    8,5
    Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 182,00 €
    182,00 €
    1 * 250 mL
       182,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61360.AK
    Physical form
    CAS No.
    MDL No.
    Appearance
    Storage temperature
  • J61360.AP
    HEPES buffer solution
    HEPES buffer solution
    1 mol/L
    8,5
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 264,00 €
    264,00 €
    1 * 500 mL
       264,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J61360.AP
    Physical form
    CAS No.
    MDL No.
    Appearance
    Storage temperature
  • J60440.AK
    HEPES buffer solution
    HEPES buffer solution
    1 mol/L
    9,0
    Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 156,00 €
    156,00 €
    1 * 250 mL
       156,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60440.AK
    Physical form
    Liquid
    CAS No.
    7365-45-9
    MDL No.
    MFCD00006158
    Appearance
    Storage temperature
  • J60440.AP
    HEPES buffer solution
    HEPES buffer solution
    1 mol/L
    9,0
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 259,00 €
    259,00 €
    1 * 500 mL
       259,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60440.AP
    Physical form
    Liquid
    CAS No.
    7365-45-9
    MDL No.
    MFCD00006158
    Appearance
    Storage temperature