Order Entry
Belgium
ContactUsLinkComponent
Buffer, HEPES solution pH 7,5 (1 mol/L)
Buffer, HEPES solution pH 7,5 (1 mol/L)
Catalog # A6916.0250
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, HEPES solution pH 7,5 (1 mol/L)
Catalog # A6916.0250
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for liquids
  • Pk:
    250 mL
  • Description:
    HEPES buffer solution pH 7,5 (1 mol/l)
  • Storage temperature:
    Ambient
  • Formulation:
    HEPES: 238.31 g/L (1 M)
  • pH:
    7.5 ± 0.2 (20°C; adjusted with NaOH)

Specifications

About this item