Order Entry
Belgium
ContactUsLinkComponent
HEPES, 0.5 M buffer solutions
HEPES, 0.5 M buffer solutions
Certificates
HEPES, 0.5 M buffer solutions
Certificates

Specifications

  • CAS No.:
    7365-45-9
  • MDL No.:
    MFCD00006158
  • Physical form:
    Liquid

Specifications

Frequently Bought Together