Order Entry
Belgium
ContactUsLinkComponent
Buffers, BIS-TRIS propane, 0,2 M solutions
Buffers, BIS-TRIS propane, 0,2 M solutions
Certificates
Buffers, BIS-TRIS propane, 0,2 M solutions
Certificates

Specifications

  • CAS No.:
    64431-96-5
  • MDL No.:
    MFCD00004689
  • Physical form:
    Liquid

Specifications

Frequently Bought Together