Sodium-D(+)-glucuronate monohydrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: D-Glucuronic acid sodium salt hydrate, D-Glucuronic acid sodium salt monohydrate, D(+)-Glucuronic acid, sodium salt, monohydrate, Sodium (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoate hydrate
CAS Number:14984-34-0
:207300-70-7
Formula:C₆H₁₀O₇·1H₂O
MDL Number:MFCD00151051
:234.14 g/mol (216.14 anhy)
Melting Pt:134…140 °C (dec.)
Storage Temperature:Ambient
EINECS:239-065-7
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Water:6.7 to 8.7 % (K.F.)
- Specific optical rotation:+20° to +22° (20°C, 589 nm) (c=2, H2O)
- Titration with HClO4:98.5 to 101.5 %
Specification Test Results
Frequently Bought Together