
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex adduct ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:95464-05-4
:
Formula:C₃₅H₃₀Cl₄FeP₂Pd
MDL Number:MFCD00015757
:816.64 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together