ContactUsLinkComponent
Membrane filters, hydrophobic edge
Membrane filters, hydrophobic edge
Supplier:  Merck
Certificates
undefined
Membrane filters, hydrophobic edge
Supplier:  Merck
Specifications
  • Catalog No:
  • MILXGSEP047S0
  • MILXHAEG047AW
  • MILFSMEP0470W
  • Type:
  • GSEP
  • HAEG, gridded
  • SMEP
  • Pore size:
  • 0,22 µm
  • 0,45 µm
  • 5,00 µm
  • Thickness:
  • -
  • -
  • 135 µm
  • Water flow rate:
  • -
  • -
  • 580 l/min/cm²
  • Airflow rate:
  • -
  • -
  • 32 l/min/cm²
  • Max. temperature:
  • 75 °C