VWR® Overalls, SMS
About this item
These durable, anti-static overalls are made of breathable, soft SMS, a spunbonded fabric with a unique tri-laminate construction, that offers a high tensile strength and toughness, plus provides a barrier to fluids and particulates.
- Elasticated hood
- Zip closure with flap
- Elasticated cuffs, ankles and waist, thumb loop
- Serged seams (stitching method)
- Low lint performance
These overalls can be used in medical, industrial, laboratory, electronic and pharmaceutical applications. Suitable for use in Critical Environments (IEST-RP-CC003.3 – Level I) and ISO Class 5 + 6 cleanrooms.
Weight: 60 g/m²
EN 340, EN 1149-5, EN 13034:2005+A1:2009 (Type 5), EN 13982-1:2004+A1:2010 (Type 6), EN 1073-2:2002; Cat. III
Certifications: EN 340, EN 1149-5, EN 13034+A1 (Type 5), EN 13982-1+A1 (Type 6), EN 1073-2; Cat. III
7 Options Available Below
Product Details & Documents
These durable, anti-static overalls are made of breathable, soft SMS, a spunbonded fabric with a unique tri-laminate construction, that offers a high tensile strength and toughness, plus provides a barrier to fluids and particulates.
- Elasticated hood
- Zip closure with flap
- Elasticated cuffs, ankles and waist, thumb loop
- Serged seams (stitching method)
- Low lint performance
These overalls can be used in medical, industrial, laboratory, electronic and pharmaceutical applications. Suitable for use in Critical Environments (IEST-RP-CC003.3 – Level I) and ISO Class 5 + 6 cleanrooms.
Weight: 60 g/m²
EN 340, EN 1149-5, EN 13034:2005+A1:2009 (Type 5), EN 13982-1:2004+A1:2010 (Type 6), EN 1073-2:2002; Cat. III
Certifications: EN 340, EN 1149-5, EN 13034+A1 (Type 5), EN 13982-1+A1 (Type 6), EN 1073-2; Cat. III
Documents
CoC Coverall CAT III Type 5,6.pdf
Déclaration de conformité
Specifications for Product Family
Specifications
- Catalog No:
- 113-8225
- 113-8226
- 113-8227
- 113-8228
- 113-8229
- 113-8230
- 113-8588
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Cleanroom|Medical|Laboratory|Clinical|Industrial
- Cleanroom|Medical|Laboratory|Clinical|Industrial
- Cleanroom|Medical|Laboratory|Clinical|Industrial
- Cleanroom|Medical|Laboratory|Clinical|Industrial
- Cleanroom|Medical|Laboratory|Clinical|Industrial
- Cleanroom|Medical|Laboratory|Clinical|Industrial
- Cleanroom|Medical|Laboratory|Clinical|Industrial
- Weight:
- 60 g/m²
- 60 g/m²
- 60 g/m²
- 60 g/m²
- 60 g/m²
- 60 g/m²
- 60 g/m²
Specifications
Product Family Options
Product Information
- ColourClothing SizeAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.113-8225Anti-static+Garment applicationCleanroom|Medical|Laboratory|Clinical|IndustrialWeight60 g/m² - Specifications:
More
Specifications
Cat. No.113-8226Anti-static+Garment applicationCleanroom|Medical|Laboratory|Clinical|IndustrialWeight60 g/m² - Specifications:
More
Specifications
Cat. No.113-8227Anti-static+Garment applicationCleanroom|Medical|Laboratory|Clinical|IndustrialWeight60 g/m² - Specifications:
More
Specifications
Cat. No.113-8228Anti-static+Garment applicationCleanroom|Medical|Laboratory|Clinical|IndustrialWeight60 g/m² - Specifications:
More
Specifications
Cat. No.113-8229Anti-static+Garment applicationCleanroom|Medical|Laboratory|Clinical|IndustrialWeight60 g/m² - Specifications:
More
Specifications
Cat. No.113-8230Anti-static+Garment applicationCleanroom|Medical|Laboratory|Clinical|IndustrialWeight60 g/m² - Specifications:
More
Specifications
Cat. No.113-8588Anti-static+Garment applicationCleanroom|Medical|Laboratory|Clinical|IndustrialWeight60 g/m²
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



