Tubing connectors, straight, reducing, PP
Catalog # SEMA0564
Supplier: SEMADENI
Specifications
- Hole Ø:8/5 mm
- Tubing int.Ø:12-16 mm/8-12 mm
- Pk:1
Specifications
About this item
Connectors, straight, conical shape and stepped, made of PP, wiith different fitting diameters on the two sides able to connect with tubing having different diameters.
- Good mechanical strength and chemical resistance
- Very good thermal resistance
- Autoclavable