ContactUsLinkComponent
Sample loops for Rheodyne® sample injectors
Sample loops for Rheodyne® sample injectors
Catalog # HYPE9055-022
Sample loops for Rheodyne® sample injectors
Catalog # HYPE9055-022
Specifications
  • Capacity:
    20 µl
  • Description:
    Sample loop, for 9010, 9725 and 9725i injectors
  • Material:
    PEEK
  • Seq:
    08
  • Pk:
    1
  • For tube I-Ø:
    0,25 mm