o-Cresolphthalein 20 g/l in ethanol 70%, VOLUSOL®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:596-27-0
:
Formula:C₂₂H₁₈O₄
MDL Number:MFCD00005912
:346.39 g/mol
Density:0.886 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-881-8
UN:1170
ADR:3,III
:GHS02
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Transition range pH 8.2 - 9.8:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC84599.180 spec_VWRC84599.260
Product specification
VWR® Indicators
Product Literature
Frequently Bought Together